

Nickel(II) hydroxide (61% Ni)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12054-48-7
:
Formula:H₂NiO₂
MDL Number:MFCD00011140
:92.71 g/mol
Melting Pt:230 °C (dec.)
Density:4.1
Storage Temperature:Ambient
EINECS:235-008-5
ADR:9,III
:GHS08
:GHS07