


Nickel(II) hydroxide for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12054-48-7
:
Formula:H₂NiO₂
MDL Number:MFCD00011140
:92.71 g/mol
Melting Pt:230 °C (dec.)
Storage Temperature:Ambient
EINECS:235-008-5
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:Green Crystalline powder
- Titration Complexometric:61.0 to 65.0 % (Ni)
- Cobalt (Co):≤8000 ppm
- Copper (Cu):≤10 ppm
- Iron (Fe):≤200 ppm
- Manganese (Mn):≤50 ppm
- Sulfur (S):≤50 ppm