Arsenazo III
Glass bottle for solids, 5 g
Catalog # MOLE13331445
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 2,7-Bis(2-arsonophenylazo)chromotropic acid, 2,7-Bis(2-arsonophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonic acid, 2,2′-(1,8-Dihydroxy-3,6-disulphonaphthylene-2,7-bisazo)bisbenzenearsonic acid, Arsenazo Ⅲ
- CAS Number:1668-00-4
- Formula:C₂₂H₁₈As₂N₄O₁₄S₂
- MDL Number:MFCD00036695
- Molecular Weight:776.38 g/mol
- EINECS:216-788-6
- UN:3465
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



