(±)-α-Lipoic acid
Glass bottle for solids, 25 g
Catalog # MOLEM68359886
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)-1,2-Dithiolane-3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid
- CAS Number:1077-28-7
- Formula:C₈H₁₄O₂S₂
- MDL Number:MFCD00005474
- Molecular Weight:206.33 g/mol
- EINECS:214-071-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



