Order Entry
Export
Orders LinkContactUsLinkComponent
(±)-α-Lipoic acid

(±)-α-Lipoic acid

Supplier:   Cayman Chemical
(±)-α-Lipoic acid
Supplier:  Cayman Chemical
Certificates
(±)-α-Lipoic acid

4 Options Available Below

Product Details & Documents
Synonyms:  5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)​-​1,2-​Dithiolane-​3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid
  • CAS Number:1077-28-7
  • Formula:C₈H₁₄O₂S₂
  • MDL Number:MFCD00005474
  • Molecular Weight:206.33 g/mol
  • EINECS:214-071-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 17,00 €
    17,00 €
    1 * 1 g
       17,00 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 57,00 €
    57,00 €
    1 * 5 g
       57,00 €

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 99,00 €
    99,00 €
    1 * 10 g
       99,00 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 214,00 €
    214,00 €
    1 * 25 g
       214,00 €