(±)-α-Lipoic acid ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 1077-28-7 Molecular Formula: C8H14O2S2 Formula Weight: 206.33 Storage Temperature: 2°C to 8°C Melting Point: 59-63° Boiling Point: 160-165° Beilstein Reference: 81853 Merck Reference: 14,9326 MDL No.: MFCD00005474
Synonyms: 5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)-1,2-Dithiolane-3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid
- CAS Number:1077-28-7
- Formula:C₈H₁₄O₂S₂
- MDL Number:MFCD00005474
- Molecular Weight:206.33 g/mol
- Merck Index:14,9326
- Boiling Pt:160…165 °C
- Melting Pt:59…63 °C
- EINECS:214-071-2
2 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 1077-28-7 Molecular Formula: C8H14O2S2 Formula Weight: 206.33 Storage Temperature: 2°C to 8°C Melting Point: 59-63° Boiling Point: 160-165° Beilstein Reference: 81853 Merck Reference: 14,9326 MDL No.: MFCD00005474
Synonyms: 5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)-1,2-Dithiolane-3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid
- CAS Number:1077-28-7
- Formula:C₈H₁₄O₂S₂
- MDL Number:MFCD00005474
- Molecular Weight:206.33 g/mol
- Merck Index:14,9326
- Boiling Pt:160…165 °C
- Melting Pt:59…63 °C
- EINECS:214-071-2



