Bis(1,5-cyclooctadiene)nickel(0), Sigma-Aldrich®
Catalog # SIAL244988-2G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:2 g
- Seq:3107
Specifications
About this item
:
:275.06 g/mol
ADR:4.1,II
:M-Clarity™ quality level = MQ200
EINECS:215-072-0
UN:1325
CAS Number:1295-35-8
Formula:C₁₆H₂₄Ni
MDL Number:MFCD00058902