Order Entry
Export
ContactUsLinkComponent
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate hydrate, Sigma-Aldrich®
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate hydrate, Sigma-Aldrich®
Catalog # SIAL683140-2G
Supplier:  Merck
CAS Number:  
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate hydrate, Sigma-Aldrich®
Catalog # SIAL683140-2G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2 g
  • Seq:
    3107

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
EINECS:460-220-1
CAS Number:207124-65-0
Formula:C₁₆H₂₄BF₄Rh·H₂O
MDL Number:MFCD00167214