Order Entry
Export
ContactUsLinkComponent
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, Sigma-Aldrich®
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, Sigma-Aldrich®
Catalog # SIAL14694-100MG
Supplier:  Merck
CAS Number:  
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, Sigma-Aldrich®
Catalog # SIAL14694-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :406.072 g/mol
EINECS:460-220-1
CAS Number:35138-22-8
Formula:C₁₆H₂₄BF₄Rh
MDL Number:MFCD00075045