Order Entry
Export
ContactUsLinkComponent
2,6-Dichloro-5-fluoronicotinonitrile, Sigma-Aldrich®
2,6-Dichloro-5-fluoronicotinonitrile, Sigma-Aldrich®
Catalog # SIAL422169-25G
Supplier:  Merck
CAS Number:  
2,6-Dichloro-5-fluoronicotinonitrile, Sigma-Aldrich®
Catalog # SIAL422169-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :190.99 g/mol
ADR:6.1,III
 :M-Clarity™ quality level = MQ100
UN:3439
CAS Number:82671-02-1
Formula:C₆HCl₂FN₂
MDL Number:MFCD01863289