2,2-Dichloro-1-methylcyclopropanecarboxylic acid, Sigma-Aldrich®
Catalog # SIAL209767-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:169.0059 g/mol
:M-Clarity™ quality level = MQ100
EINECS:215-901-6
CAS Number:1447-14-9
Formula:C₅H₆Cl₂O₂
MDL Number:MFCD00001291