Order Entry
Export
ContactUsLinkComponent
2,2-Difluorocyclopropanecarboxylic acid, Sigma-Aldrich®
2,2-Difluorocyclopropanecarboxylic acid, Sigma-Aldrich®
Catalog # SIAL684678-250MG
Supplier:  Merck
CAS Number:  
2,2-Difluorocyclopropanecarboxylic acid, Sigma-Aldrich®
Catalog # SIAL684678-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :122.07 g/mol
ADR:8,III
 :M-Clarity™ quality level = MQ100
UN:3261
CAS Number:107873-03-0
Formula:C₄H₄F₂O₂
MDL Number:MFCD04115828