
2,2-Difluorocyclopropanecarboxylic acid 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:107873-03-0
:
Formula:C₄H₄F₂O₂
MDL Number:MFCD04115828
:122.07 g/mol
Boiling Pt:70 °C (7 mmHg)
Melting Pt:55…60 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05