5,7-Diiodo-8-quinolinol, Sigma-Aldrich®
Catalog # SIALD123609-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
EINECS:201-497-9
CAS Number:83-73-8
Formula:C₉H₅I₂NO
MDL Number:MFCD00006789