Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Benzofuran-2-carboxylic acid
Benzofuran-2-carboxylic acid
Benzofuran-2-carboxylic acid
Benzofuran-2-carboxylic acid
Supplier:
CHEMPUR
Certificates
Expand image
Benzofuran-2-carboxylic acid
Supplier:
CHEMPUR
About this item
CAS Number:
496-41-3
:
Formula:
C₉H₆O₃
MDL Number:
MFCD00005848
:
162.15 g/mol
EINECS:
207-818-9