Order Entry
Export
ContactUsLinkComponent
3-(3,4-Dihydroxyphenyl)-L-alanine
3-(3,4-Dihydroxyphenyl)-L-alanine
Catalog # PROOMM0262.00
CAS Number:  
3-(3,4-Dihydroxyphenyl)-L-alanine
Catalog # PROOMM0262.00
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :197.19 g/mol
EINECS:200-445-2
CAS Number:59-92-7
Formula:C₉H₁₁NO₄
MDL Number:MFCD00002598