
3-(3,4-Dihydroxyphenyl)-L-alanine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:59-92-7
:
Formula:C₉H₁₁NO₄
MDL Number:MFCD00002598
:197.19 g/mol
Melting Pt:295 °C (dec.)
EINECS:200-445-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to cream Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 %
- Sulfated ash:≤0.1 %
- HPLC:≥98.5 %
- Titration with HClO4:≥98.5 % (On dried substance)