Order Entry
Export
ContactUsLinkComponent
1,1-Cyclohexanedicarboxylic acid, Sigma-Aldrich®
1,1-Cyclohexanedicarboxylic acid, Sigma-Aldrich®
Catalog # SIAL708216-1G
Supplier:  Merck
CAS Number:  
1,1-Cyclohexanedicarboxylic acid, Sigma-Aldrich®
Catalog # SIAL708216-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :172.18 g/mol
CAS Number:1127-08-8
Formula:C₈H₁₂O₄
MDL Number:MFCD00094668