
1,1-Cyclohexanedicarboxylic acid 95%
Catalog # H26025.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
CAS Number:1127-08-8
:
Formula:C₈H₁₂O₄
MDL Number:MFCD00094668
:172.18 g/mol
Melting Pt:∼ 175 °C (dec.)
:GHS07
Storage Temperature:Ambient