
Diethyl cyclohexane-1,1-dicarboxylate 95%
Catalog # H34412.MD
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
Specifications
About this item
:
:228.29 g/mol
Boiling Pt:106…108 °C (1 mmHg)
:GHS07
Storage Temperature:Ambient
CAS Number:1139-13-5
Formula:C₁₂H₂₀O₄
MDL Number:MFCD02093457