2,4'-Dichloroacetanilide
Catalog # MAYBSPB00454.50
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:204.0533 g/mol
CAS Number:3289-75-6
Formula:C₈H₇Cl₂NO
MDL Number:MFCD00018904