4,5-Diaminopyrimidine, Sigma-Aldrich®
Catalog # SIALD24501-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:110.1200 g/mol
:M-Clarity™ quality level = MQ100
EINECS:237-339-0
CAS Number:13754-19-3
Formula:C₄H₆N₄
MDL Number:MFCD00006113