ContactUsLinkComponent
General purpose gloves, ActivArmr® 52-502
General purpose gloves, ActivArmr® 52-502
Supplier:  Ansell
Certificates
undefined
General purpose gloves, ActivArmr® 52-502
Supplier:  Ansell
Specifications
  • Catalog No:
  • ANSE52-547/10
  • ANSE52-547/9
  • Model:
  • 52-547
  • 52-547
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Liner material:
  • Cotton
  • Cotton
  • Finish:
  • Palm impregnated
  • Palm impregnated
  • Cuff style:
  • Safety cuff
  • Safety cuff
  • EN norm:
  • EN 388:2016 (3111B)
  • EN 388:2016 (3111B)
Specifications