Order Entry
Export
ContactUsLinkComponent
(+)-Dehydroabietylamine 60%, Technical Grade, Sigma-Aldrich®
(+)-Dehydroabietylamine 60%, Technical Grade, Sigma-Aldrich®
Catalog # SIAL105198-100G
Supplier:  Merck
CAS Number:  
(+)-Dehydroabietylamine 60%, Technical Grade, Sigma-Aldrich®
Catalog # SIAL105198-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :285.47 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:215-899-7
CAS Number:1446-61-3
Formula:C₂₀H₃₁N
MDL Number:MFCD00213430