0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
(±)-1,1'-Bi-2-naphthol
(±)-1,1'-Bi-2-naphthol, Sigma-Aldrich®
(±)-1,1'-Bi-2-naphthol, Sigma-Aldrich®
(±)-1,1'-Bi-2-naphthol, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
(±)-1,1'-Bi-2-naphthol, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
602-09-5
:
Formula:
C₂₀H₁₄O₂
MDL Number:
MFCD00004068
:
286.33 g/mol
ADR:
6.1,III
:
M-Clarity™ quality level = MQ200
EINECS:
210-014-0
UN:
2811