Dodecenylsuccinic anhydride (mixture of isomers), Sigma-Aldrich®
Catalog # SIAL443271-1L
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:266.38 g/mol
:M-Clarity™ quality level = MQ100
EINECS:246-917-1
CAS Number:25377-73-5
:26544-38-7
Formula:C₁₆H₂₆O₃
MDL Number:MFCD00036197