(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, Sigma-Aldrich®
Catalog # SIAL693049-500MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:678.78 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:99646-28-3
Formula:C₄₈H₄₀P₂
MDL Number:MFCD01311709