Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Dihydroxyacetone dimer
1,3-Dihydroxyacetone dimer, Sigma-Aldrich®
1,3-Dihydroxyacetone dimer, Sigma-Aldrich®
1,3-Dihydroxyacetone dimer, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
1,3-Dihydroxyacetone dimer, Sigma-Aldrich®
Supplier:
Merck
Certificates
About this item
CAS Number:
62147-49-3
Formula:
C₆H₁₂O₆
MDL Number:
MFCD00051019
:
180.16 g/mol
:
M-Clarity™ quality level = MQ100
Related Information
Documentation
Certificates
Search by Lot Number
Documentation