2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid)
Catalog # MOLE16238403-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:144.13 g/mol
EINECS:217-992-8
CAS Number:2033-24-1
Formula:C₆H₈O₄
MDL Number:MFCD00006638