Pesticide reference standard, 2,3,5-Trichlorophenyl acetate 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL17775200IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:61925-88-0
- Description:2,3,5-Trichlorophenyl acetate 10 µg/ml in isooctane
- Formula:C₈H₅Cl₃O₂
- Description:2,3,5-Trichlorophenyl acetate