2,6-Dichlorobenzylideneacetone
Catalog # MAYBCD09843.50
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:215.08 g/mol
CAS Number:81559-89-9
Formula:C₁₀H₈Cl₂O
MDL Number:MFCD00051609