0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2,4-Dichlorobenzylideneacetone
2,4-Dichlorobenzylideneacetone ≥97%
2,4-Dichlorobenzylideneacetone ≥97%
2,4-Dichlorobenzylideneacetone ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
2,4-Dichlorobenzylideneacetone ≥97%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
61888-78-6
:
Formula:
C₁₀H₈Cl₂O
MDL Number:
MFCD00052852
:
215.08 g/mol
Melting Pt:
83…85 °C
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFAB25270
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation