Order Entry
Export
Orders LinkContactUsLinkComponent
Progesterone 98%

Progesterone 98%

Progesterone 98%
Certificates
Progesterone 98%

3 Options Available Below

Product Details & Documents
  • CAS Number:57-83-0
  • Formula:C₂₁H₃₀O₂
  • MDL Number:MFCD00003658
  • Melting Pt:126…131 °C
  • Storage Temperature:Ambient
  • EINECS:200-350-6

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    126 to 131 °C
  • Loss on drying:
    ≤0.5 % (500 mg, 105 °C, 2 hrs)
  • HPLC:
    ≥97.5 %
  • Specific optical rotation:
    +186° to +194° (20 °C, 589 nm) (c=1, ethanol) (on dried substance)
Product Family Options

Product Information