3,4-(Methylenedioxy)phenylacetonitrile, Sigma-Aldrich®
Catalog # SIAL145300-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:161.16 g/mol
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:224-655-9
UN:3439
CAS Number:4439-02-5
Formula:C₉H₇NO₂
MDL Number:MFCD00005835