Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Nitro-1,3-benzodioxole
5-Nitro-1,3-benzodioxole, Sigma-Aldrich®
5-Nitro-1,3-benzodioxole, Sigma-Aldrich®
5-Nitro-1,3-benzodioxole, Sigma-Aldrich®
SIAL161500-5G
:
Merck
:
5-Nitro-1,3-benzodioxole, Sigma-Aldrich®
SIAL161500-5G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
167.12 g/mol
:
M-Clarity™ quality level = MQ100
:
220-055-6
:
2620-44-2
:
C₇H₅NO₄
:
MFCD00005824