2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde, Sigma-Aldrich®
Catalog # SIAL594423-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:186.11 g/mol
:M-Clarity™ quality level = MQ200
CAS Number:656-42-8
Formula:C₈H₄F₂O₃
MDL Number:MFCD00792420