Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloro-1,3-benzodioxole
5-Chloro-1,3-benzodioxole ≥98%
5-Chloro-1,3-benzodioxole ≥98%
:
Thermo Fisher Scientific
5-Chloro-1,3-benzodioxole ≥98%
:
Thermo Fisher Scientific
:
7228-38-8
:
:
C₇H₅ClO₂
:
MFCD00010842
:
156.57 g/mol
:
185…187 °C
:
1.340
:
93 °C (201 °F)
:
GHS07
:
Ambient
msds_ALFAB22534
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years