4,4'-Methylenebis(2,6-diethylaniline), Sigma-Aldrich®
Catalog # SIAL360783-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:310.4763 g/mol
EINECS:237-185-4
REACH:01-0000015192-78
CAS Number:13680-35-8
Formula:C₂₁H₃₀N₂
MDL Number:MFCD00071552