5,8-Dihydroxy-1,4-naphthoquinone, Sigma-Aldrich®
Catalog # SIAL388513-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:190.16 g/mol
:M-Clarity™ quality level = MQ100
EINECS:207-495-4
CAS Number:475-38-7
Formula:C₁₀H₆O₄
MDL Number:MFCD00001685