Invertase
Catalog # SIALI0408-500UG
Supplier: Merck
CAS Number:
Specifications
- Pk:500 µG
- CAS No.:9001-57-4
- Form:Lyophilised powder
- Enzyme name:Invertase
Specifications
About this item
Invertase glycoprotein standard.
- Used to generate N-linked glycan library
- As a negative control to study the binding of lectins to high mannose structures
- For sample pre-treatment in proteomic analyses to study drug-induced toxic epidermal necrolysis
The invertase glycoprotein standard can be used to demonstrate N-glycosylation using PNGase F with both in-solution and in-gel procedures. The extent of deglycosylation can be assessed by mobility shift on SDS-PAGE gels.
Used in the production of confectionary foods and artificial honey.