Order Entry
Export
ContactUsLinkComponent
 
 
5-Bromo-1,3-benzodioxole 97%
 
undefined
 
5-Bromo-1,3-benzodioxole 97%

 

  • Appearance:
    Clear yellow to light brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5820 to 1.5850 (20°C, 589 nm)
  • GC:
    ≥96.0 %