2-Chlorophenyl phosphorodichloridate, Sigma-Aldrich®
Catalog # SIAL235237-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:245.43 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:239-129-4
UN:3265
CAS Number:15074-54-1
Formula:C₆H₄Cl₃O₂P
MDL Number:MFCD00009704