Organic reference standard, 2,4,6-Tribromobiphenyl (PBB No. 30) 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL21003000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:59080-33-0
- Description:2,4,6-Tribromobiphenyl (PBB No. 30) 10 µg/ml in cyclohexane
- Formula:C₁₂H₇Br₃
- Description:2,4,6-Tribromobiphenyl (PBB No. 30)