Organic reference standard, 6-Methylchrysene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
EHERL20890000AL
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:1705-85-7
- Description:6-Methylchrysene 10 µg/ml in acetonitrile
- Formula:C₁₉H₁₄
- Description:6-Methylchrysene