4,4'-Dihydroxydiphenylmethane, Sigma-Aldrich®
Catalog # SIALB47006-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
:200.24 g/mol
EINECS:210-658-2
CAS Number:620-92-8
Formula:C₁₃H₁₂O₂
MDL Number:MFCD00002385