4,4'-Dihydroxydiphenylmethane
Catalog # EHERC10655800
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:200.24 g/mol
EINECS:210-658-2
CAS Number:620-92-8
Formula:C₁₃H₁₂O₂
MDL Number:MFCD00002385