Order Entry
Export
Orders LinkContactUsLinkComponent
2,4-Dichlorophenyl acetate

2,4-Dichlorophenyl acetate

Supplier:   CHEMPUR
2,4-Dichlorophenyl acetate
Supplier:  CHEMPUR
Certificates
2,4-Dichlorophenyl acetate

2 Options Available Below

Product Details & Documents
  • CAS Number:6341-97-5
  • Formula:C₈H₆Cl₂O₂
  • Molecular Weight:205.0380 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

This product has been blocked by your organization. Please contact your purchasing department for more information