Order Entry
Export
ContactUsLinkComponent
4,4'-Dibromooctafluorobiphenyl (DBOB)
4,4'-Dibromooctafluorobiphenyl (DBOB)
Catalog # EHERC12240600
Supplier:  EHRENSTORFER
CAS Number:  
4,4'-Dibromooctafluorobiphenyl (DBOB)
Catalog # EHERC12240600
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :455.94 g/mol
EINECS:233-847-1
CAS Number:10386-84-2
Formula:C₁₂Br₂F₈
MDL Number:MFCD00000310