Order Entry
Export
Orders LinkContactUsLinkComponent
Vinylferrocene

Vinylferrocene

Glass bottle for solids, 1 g
Catalog # 17450.03
Vinylferrocene
Catalog # 17450.03
CAS Number:  1271-51-8
Vinylferrocene
Vinylferrocene

Glass bottle for solids, 1 g

CAS Number:  1271-51-8
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents

CAS No.: 1271-51-8 Molecular Formula: C12H12Fe Formula Weight: 212.07 Flash Point: 62°(144°F) Physical Form: Powder Melting Point: 52-55° Boiling Point: 80-85°/0.7mm MDL No.: MFCD00001434

  • CAS Number:1271-51-8
  • Formula:C₁₂H₁₂Fe
  • MDL Number:MFCD00001434
  • Molecular Weight:212.07 g/mol
  • Boiling Pt:80…85 °C (0.7 mmHg)
  • Melting Pt:52…55 °C
  • Flash Pt:62 °C (144 °F)
  • Storage Temperature:Ambient
  • EINECS:215-041-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications