Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Benzodioxole-4-carboxylic acid
1,3-Benzodioxole-4-carboxylic acid
1,3-Benzodioxole-4-carboxylic acid
MAYBCC01701DA
:
Thermo Scientific
:
1,3-Benzodioxole-4-carboxylic acid
MAYBCC01701DA
:
Thermo Scientific
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
166.1308 g/mol
:
5768-39-8
:
C₈H₆O₄
:
MFCD01076411